missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSM Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | OSM |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825835
|
Proteintech
27792-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815835
|
Proteintech
27792-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Oncostatin M is a member of a cytokine family that includes leukemia-inhibitory factor, granulocyte colony-stimulating factor, and interleukin 6. This gene encodes a growth regulator which inhibits the proliferation of a number of tumor cell lines. It regulates cytokine production, including IL-6, G-CSF and GM-CSF from endothelial cells.Specifications
OSM | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human | |
P13725, Q65Z15 | |
289747, 5008 | |
OSM Fusion Protein Ag27093 | |
Primary | |
-20°C | |
OSM |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
oncostatin M, OSM | |
Osm | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |