missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSMR Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10982-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the type I cytokine receptor family. The encoded protein heterodimerizes with interleukin 6 signal transducer to form the type II oncostatin M receptor and with interleukin 31 receptor A to form the interleukin 31 receptor, and thus transduces oncostatin M and interleukin 31 induced signaling events. Mutations in this gene have been associated with familial primary localized cutaneous amyloidosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
OSMR | |
Polyclonal | |
Unconjugated | |
OSMR | |
IL 31 receptor subunit beta, IL 31R beta, IL 31R subunit beta, IL 31RB, oncostatin M receptor, OSMR, OSMR beta, OSMRB, OSMRbeta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9180 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99650 | |
OSMR | |
OSMR Fusion Protein Ag1426 | |
20 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |