Läs mer
Ospemifene, TRC
Produktkod. 30136263
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136263 1 g
30136891 500 mg
17914015 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136263 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO7030001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 128607-22-7
Molekylformel C24 H23 Cl O2
Molekylvikt (g/mol) 378.89
Synonym Ethanol, 2-[4-[(1Z)-4-chloro-1,2-diphenyl-1-buten-1-yl]phenoxy]-, Ethanol, 2-[4-(4-chloro-1,2-diphenyl-1-butenyl)phenoxy]-, (Z)-, Ethanol, 2-[4-[(1Z)-4-chloro-1,2-diphenyl-1-butenyl]phenoxy]- (9CI), 2-[4-[(1Z)-4-Chloro-1,2-diphenyl-1-buten-1-yl]phenoxy]ethanol, FC 1271a, Fc 1271, Ospemifene, Osphena
IUPAC-namn 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]ethanol
LEDER OCCOc1ccc(cc1)\C(=C(\CCCl)/c2ccccc2)\c3ccccc3
Specifikationer
Kemiskt namn eller material Ospemifene
Kvantitet 1 g
InChI formel InChI=1S/C24H23ClO2/c25-16-15-23(19-7-3-1-4-8-19)24(20-9-5-2-6-10-20)21-11-13-22(14-12-21)27-18-17-26/h1-14,26H,15-18H2/b24-23-
Formel vikt 378.1387