missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteocalcin Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Osteocalcin |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839804
|
Proteintech
20277-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829804
|
Proteintech
20277-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody is specific to mouse Bglap1.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.Specifications
Osteocalcin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse | |
P86546 | |
12096 | |
Peptide | |
Primary | |
-20°C | |
Bglap |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Bglap, Bglap1, mOC A, OC, OG1, Osteocalcin | |
Bglap | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |