missing translation for 'onlineSavingsMsg'
Learn More

OSTF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10671-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827203

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Osteoclast-stimulating factor-1 is an intracellular protein produced by osteoclasts that indirectly induces osteoclast formation and bone resorption.
Specifications

Specifications

OSTF1
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q62422, Q6P686, Q92882
OSF, OSTF1, SH3P2
Rabbit
IgG
150 μL
-20°C
Primary
20409, 259275, 26578
Immunohistochemistry (Paraffin)
Unconjugated
OSTF1
Liquid
Ostf1
OSTF1 Fusion Protein Ag1072
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers