missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | OSTM1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823354
|
Proteintech
14621-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16813354
|
Proteintech
14621-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile maligt form of osteopetrosis.Specifications
OSTM1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q86WC4, Q8BGT0 | |
14628, 28962 | |
OSTM1 Fusion Protein Ag6254 | |
Primary | |
-20°C | |
OSTM1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GIPN, GL, HSPC019, OPTB5, OSTM1, UNQ6098/PRO21201 | |
OSTM1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |