Learn More
OTUD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558418
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SSAAEPVIVS RSDPRDEKLA LYLAEVEKQD KYLRQRNKYR FHIIPDGNCL YRAVSKTVYG DQSLHRELRE QTVHYIADHL DHFSPLIEGD VGEFIIAAAQ D Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
OTUD1 Polyclonal antibody specifically detects OTUD1 in Human samples. It is validated for Immunohistochemistry.Specifications
OTUD1 | |
Polyclonal | |
Unconjugated | |
OTUD1 | |
DUBA7; DUBA-7; OTDC1; OTU deubiquitinase 1; OTU domain containing 1; OTU domain-containing protein 1; OTUD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VV17 | |
OTUD1 | |
Recombinant protein corresponding to Human OTUD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only