Läs mer
Ouabain, TRC
Produktkod. 18090920
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18090920 50 mg
18033130 1 mg
18073720 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18090920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO71450550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 630-60-4
Molekylformel C29 H44 O12
Molekylvikt (g/mol) 584.65
Synonym Card-20(22)-enolide, 3-[(6-deoxy-α-L-mannopyranosyl)oxy]-1,5,11,14,19-pentahydroxy-, (1β,3β,5β,11α)-, Ouabain (8CI), (1β,3β,5β,11α)-3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,5,11,14,19-pentahydroxycard-20(22)-enolide, Acocantherin, Astrobain, Gratibain, Gratus strophanthin, Kombetin, NSC 25485, Ouabaine, Purostrophan, Rectobaina, Solufantina, Strodival, Strophalen, Strophoperm, Strophosan, Uabaina, Uabanin, g-Strophanthin, g-Strophicor
IUPAC-namn 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
LEDER C[C@@H]1O[C@@H](O[C@H]2C[C@@H](O)[C@]3(CO)[C@H]4[C@H](O)C[C@]5(C)[C@H](CC[C@]5(O)[C@@H]4CC[C@]3(O)C2)C6=CC(=O)OC6)[C@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material G-Strophanthin
Kvantitet 50 mg
InChI formel InChI=1S/C29H44O12/c1-13-22(34)23(35)24(36)25(40-13)41-15-8-19(32)28(12-30)21-17(3-5-27(28,37)9-15)29(38)6-4-16(14-7-20(33)39-11-14)26(29,2)10-18(21)31/h7,13,15-19,21-25,30-32,34-38H,3-6,8-12H2,1-2H3/t13-,15-,16+,17+,18+,19+,21+,22-,23+,24+,25-,26+,27-,28+,29-/m0/s1
Formel vikt 584.2833