Learn More
VistaLab™ Technologies Ovation™ Pipet Accessories: Reconditioning Nozzle Kit
For Ovation E8 & E12 Multichannel Pipets
Brand:  VistaLab™ Technologies 9060-3011
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11976042
  • 3328.26 SEK / 1 set
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Contains 2 nozzles, 24 nozzle seals, 24 filters & nozzle removal tool
Specifications
Specifications
Large Nozzle Reconditioning Kit
25 to 850 μL, 25 to 1250 μL
2 Nozzles, 24 seals, 24 filters and tool
2 in.
5 in.
Non-sterile
25 to 850 μL, 25 to 1250 μL Ovation™ BioNatural™ Multichannel Pipette
1 Ea.
2 in.