Learn More
Chemsplash OverBoot with White PVC Sole, Elasticated Ankle and Top
Product Code. 30058443
Change view
Click to view available options
Size:
X-Large
2X-Large
Unit Size:
200 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058443 X-Large 200 pairs
30058293 2X-Large 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30058443 Supplier Chemsplash Supplier No. 2565.400.007
  • ONLINE EXCLUSIVE PRICE
    7950.23 SEK
    6753.79 SEK / 200 par
    Save 1196.44 SEK
    15% Av
Beräknad leverans: 18-12-2026
The Non-Slip Overboot is elasticated at the calf and ankle and has a non-slip white PVC sole.
Chemsplash Non-Slip Overboot
  • Chemsplash 55GSM Type PB 6B Material
  • Elasticated Top and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • White PVC Sole
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Non-Slip Overboot is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust. This overboot is elasticated at the calf and ankle and has a non-slip white PVC sole. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Size X-Large
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Anti-slip
Resistance Liquid
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100-1000 (ISO 5-6)