Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs
Eliminate comb assembly with these heavy-duty, one-piece, standard seven-tooth combs.
590.00 SEK - 615.00 SEK
Specifications
Description | Standard comb |
---|---|
For Use With (Equipment) | Owl™ C2-S Micro Electrophoresis System |
Product Line | Owl |
Type | Comb |
Teeth Count | 7 |
Product Code | Brand | Description | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | |||
12404587
|
Thermo Scientific™
C2-7C |
Standard comb | 7 | 1.0 mm |
590.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
12414587
|
Thermo Scientific™
C2-7D |
Standard comb | 7 | 1.5 mm |
615.00 SEK
Each |
In Stock
Log in to see stock. |
|||
Description
Eliminate comb assembly with heavy-duty, one-piece Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs. These standard seven-tooth combs are available in two thicknesses (1.0 and 1.5mm) and are for use with the Owl™ C2-S Micro Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.Specifications
Standard comb | |
Owl | |
7 |
Owl™ C2-S Micro Electrophoresis System | |
Comb |