Learn More
Thermo Scientific™ Owl™ D3-14 System Replacement Parts
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts including UVT gel tray, gaskets and external caster.
206.00 SEK - 1645.00 SEK
Specifications
For Use With (Equipment) | Owl™ D3 Series Horizontal Electrophoresis System |
---|---|
Product Line | Owl |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11912975
|
Thermo Scientific™
D3-CST-14 |
External caster (no trays/combs) |
1645.00 SEK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
11977515
|
Thermo Scientific™
D3-GK |
Replacement Gaskets (1 pair) |
206.00 SEK
Pack of 2 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
11922975
|
Thermo Scientific™
D3-UVT-14 |
EasyCast™ gasketed UVT Gel Tray |
1505.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
Descrizione
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned to enable up to 12 sample sets on the same gel
Specifica
Owl™ D3 Series Horizontal Electrophoresis System | |
Owl |