Läs mer
Oxalic Acid-(1,2-13C2), TRC
Produktkod. 30011054
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011054 25 mg
30011536 5 mg
30011678 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011054 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84512325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62654-02-8
Molekylformel 13C2H2O4
Molekylvikt (g/mol) 92.02
Synonym Ethanedioic Acid-(1,2-13C2), Aktisal-(1,2-13C2), Aquisal-(1,2-13C2), DeerClean-(1,2-13C2), NSC 132055-(1,2-13C2), NSC 151956-(1,2-13C2), NSC 62774-(1,2-13C2), NSC 76990-(1,2-13C2), Ultraplast Activate S 52-(1,2-13C2);
LEDER O=[13C](O)[13C](O)=O
Specifikationer
Kemiskt namn eller material Oxalic Acid-(1,2-13C2)
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)/i1+1,2+1
Formel vikt 92.002
Procent renhet >95%