Läs mer
Oxalic Acid Ethyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Oxalic Acid Ethyl Ester |
| CAS | 617-37-8 |
| Molekylformel | C4H6O4 |
| Kvantitet | 5 g |
| Synonym | Ethanedioic Acid Monoethyl Ester, Oxalic Acid Ethyl Ester, Oxalic Acid Monoethyl Ester, Ethyl Hydrogen Oxalate, Monoethyl Oxalate |
| InChI formel | InChI=1S/C4H6O4/c1-2-8-4(7)3(5)6/h2H2,1H3,(H,5,6) |
| LEDER | O=C(C(OCC)=O)O |
| Molekylvikt (g/mol) | 118.09 |
| Formel vikt | 118.03 |