Läs mer
Oxaloacetic Acid-13C4, TRC
Produktkod. 30134766
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134766 2.5 mg 2.5mg
30133686 5 mg -
17932385 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8450322.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161096-82-8
Molekylformel 13C4H4O5
Molekylvikt (g/mol) 136.04
Synonym Oxobutanedioic Acid-13C4, Oxalacetic Acid-13C4, 2-Ketosuccinic Acid-13C4, 2-Oxobutanedioic Acid-13C4, 2-Oxosuccinic Acid-13C4, Ketosuccinic Acid-13C4, NSC 284205-13C4, NSC 77688-13C4, OAA-13C4, Oxaloacetic Acid-13C4, Oxaloethanoic Acid-13C4, Oxosuccinic Acid-13C4, α-Ketosuccinic Acid-13C4
LEDER O=[13C]([13C]([13CH2][13C](O)=O)=O)O
Specifikationer
Kemiskt namn eller material Oxaloacetic Acid-13C4
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)/i1+1,2+1,3+1,4+1
Formel vikt 136.0193
Procent renhet >95%