Läs mer
Oxamflatin Ethyl Ester, TRC
Produktkod. 30415050
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30415050 100 mg
1 options
Produktkod. 30415050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO845095100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxamflatin Ethyl Ester
CAS 342373-23-3
Molekylformel C19H17NO4S
Kvantitet 100 mg
Synonym 5-[3-[(Phenylsulfonyl)amino]phenyl]-2-penten-4-ynoic Acid Ethyl Ester
InChI formel InChI=1S/C19H17NO4S/c1-2-24-19(21)14-7-6-9-16-10-8-11-17(15-16)20-25(22,23)18-12-4-3-5-13-18/h3-5,7-8,10-15,20H,2H2,1H3/b14-7+
LEDER CCOC(=O)\C=C\C#Cc1cccc(NS(=O)(=O)c2ccccc2)c1
IUPAC-namn ethyl (E)-5-[3-(benzenesulfonamido)phenyl]pent-2-en-4-ynoate
Molekylvikt (g/mol) 355.41
Formel vikt 355.0878