Läs mer
Oxazolidine-4-carboxylic acid hydrochloride, TRC
Produktkod. 30414861
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30414861 50 mg
1 options
Produktkod. 30414861 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO99105050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4S)-1,3-oxazolidine-4-carboxylic acid hydrochloride
CAS 162285-30-5
Molekylformel C4H7NO3.HCl
Kvantitet 50 mg
Synonym 4-Oxazolidinecarboxylic acid','hydrochloride (1:1) (ACI), oxazolidine-4-carboxylic acid hydrochloride
InChI formel InChI=1S/C4H7NO3.ClH/c6-4(7)3-1-8-2-5-3;/h3,5H,1-2H2,(H,6,7);1H
LEDER Cl.O=C(O)C1NCOC1
IUPAC-namn oxazolidine-4-carboxylic acid hydrochloride
Molekylvikt (g/mol) 153.56
Formel vikt 153.02