Läs mer
Oxazolidine Cabazitaxel, TRC
Produktkod. 30426672
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30426672 500 mg
1 options
Produktkod. 30426672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO846530500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxazolidine Cabazitaxel
CAS 1373171-12-0
Molekylformel C48 H61 N O14
Kvantitet 500 mg
Synonym 3,5-Oxazolidinedicarboxylic acid','2,2-dimethyl-4-phenyl-','5-[(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl] 3-(1,1-dimethylethyl) ester','(4S,5R)- (ACI), 5-[(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl] 3-(1,1-dimethylethyl) (4S,5R)-2,2-dimethyl-4-phenyl-3,5-oxazolidinedicarboxylate (ACI), 4-Acetoxy-2a-benzoyloxy-5β-20-epoxy-1-hydroxy-9-oxo-7β,10β-dimethoxy-11-taxen-13-yl (4S,5R)-3-(tert-butylcarbonyl)-2,2-dimethyl-4-phenyl-5-oxazolidinecarboxylate
InChI formel InChI=1S/C48H61NO14/c1-26-30(59-41(53)36-34(28-19-15-13-16-20-28)49(45(8,9)62-36)42(54)63-43(3,4)5)24-48(55)39(60-40(52)29-21-17-14-18-22-29)37-46(10,38(51)35(57-12)33(26)44(48,6)7)31(56-11)23-32-47(37,25-58-32)61-27(2)50/h13-22,30-32,34-37,39,55H,23-25H2,1-12H3/t30-,31-,32+,34-,35+,36+,37-,39-,46+,47-,48+/m0/s1
LEDER CO[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@@H]6OC(C)(C)N([C@H]6c7ccccc7)C(=O)OC(C)(C)C)C(=C([C@@H](OC)C(=O)[C@]13C)C5(C)C)C
Molekylvikt (g/mol) 875.996
Formel vikt 875.409