Läs mer
Oxethazaine, MedChemExpress
Produktkod. 16414517
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16414517 50 mg
1 options
Produktkod. 16414517 Leverantör Medchem Express Leverantörsnummer HYB0955
MedChemExpress Oxethazaine (Oxetacaine), a precursor of phentermine acidic, is an acid-resistent and orally active analgesic agent. Oxethazaine (Oxetacaine) has the potential for the relief of pain associated with peptic ulcer disease or esophagitis.
Oxethazaine (Oxetacaine), a precursor of phentermine acidic, is an acid-resistent and orally active analgesic agent. Oxethazaine (Oxetacaine) has the potential for the relief of pain associated with peptic ulcer disease or esophagitis.
Specifikationer
Kemiskt namn eller material Oxethazaine
Fysisk form Solid
CAS 126-27-2
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C28H41N3O3
För användning med (applikation) Neuroscience-Neuromodulation
Synonym Oxetacaine
Löslighetsinformation DMSO : ≥ 50 mg/mL (106.92 mM)
LEDER OCCN(CC(N(C(C)(C)CC1=CC=CC=C1)C)=O)CC(N(C(C)(C)CC2=CC=CC=C2)C)=O
Molekylvikt (g/mol) 467.64
Formel vikt 467.64
Procent renhet 99.07%