missing translation for 'onlineSavingsMsg'
Läs mer

Oxiconazole Nitrate, TRC

Produktkod. 30428457
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428457 5 mg
30428112 100 mg
30411133 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30428457 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8468005MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 64211-46-7
Molekylformel C18 H13 Cl4 N3 O . H N O3
Molekylvikt (g/mol) 492.14
Synonym Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (1Z)-, mononitrate (1:1), Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (1Z)-, mononitrate (9CI), Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (Z)-, mononitrate, Gyno-Myfungar, Myfungar, Oceral, Oxiconazole nitrate, Oxistat, Ro 13-8996, ST 813, Scorba LX, Sgd 301-76
IUPAC-namn (Z)-1-(2,4-dichlorophenyl)-N-[(2,4-dichlorophenyl)methoxy]-2-imidazol-1-ylethanimine;nitric acid
LEDER O[N+](=O)[O-].Clc1ccc(CO\N=C(/Cn2ccnc2)\c3ccc(Cl)cc3Cl)c(Cl)c1

Specifikationer

Kemiskt namn eller material Oxiconazole Nitrate
Kvantitet 5 mg
InChI formel InChI=1S/C18H13Cl4N3O.HNO3/c19-13-2-1-12(16(21)7-13)10-26-24-18(9-25-6-5-23-11-25)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11H,9-10H2;(H,2,3,4)/b24-18+;
Formel vikt 489.9769
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.