Läs mer
Novus Biologicals™ Oxidized LDL Antibody (Okayama U. patent anti-oxLDL) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28420-1mg
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Oxidized LDL Monoclonal antibody specifically detects Oxidized LDL in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifikationer
| Oxidized LDL | |
| Monoclonal | |
| Unconjugated | |
| Oxidized LDL, Oxidized Low Density Lipoprotein, oxLDL | |
| oxLDL | |
| 1 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Okayama U. patent anti-oxLDL | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Human | |
| Protein A purified | |
| RUO | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |