Läs mer
Oxirane-2-carboxylic Acid, TRC
Produktkod. 30412063
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30412063 10 mg
1 options
Produktkod. 30412063 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84689010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxirane-2-carboxylic Acid
CAS 503-11-7
Molekylformel C3 H4 O3
Kvantitet 10 mg
Synonym Glycidic Acid, Oxiranecarboxylic Acid, 2,3-Epoxypropionic Acid, Epihydrinic Acid, Ethylene Oxide-carboxylic Acid, 2,3-Epoxy-propanoic Acid
InChI formel InChI=1S/C3H4O3/c4-3(5)2-1-6-2/h2H,1H2,(H,4,5)
LEDER OC(=O)C1CO1
IUPAC-namn oxirane-2-carboxylic acid
Molekylvikt (g/mol) 88.062
Formel vikt 88.016