Läs mer
Oxo-Pemetrexed acid, TRC
Produktkod. 30318489
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30318489 5 mg
1 options
Produktkod. 30318489 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6299105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2-(2-Amino-4,5,6,7-tetrahydro-4,6-dioxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid
CAS 193265-47-3
Molekylformel C15H14N4O4
Kvantitet 5 mg
Synonym 4-[2-(2-Amino-4,5,6,7-tetrahydro-4,6-dioxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid, Oxo-Pemetrexed acid, Benzoic acid, 4-[2-(2-amino-4,5,6,7-tetrahydro-4,6-dioxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]- (ACI), 4-[2-(2-Amino-4,5,6,7-tetrahydro-4,6-dioxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid (ACI), Benzoic acid, 4-[2-(2-amino-4,5,6,7-tetrahydro-4,6-dioxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]- (9CI), 4-[2-(2-Amino-4,6-dioxo-5,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid
InChI formel InChI=1S/C15H14N4O4/c16-15-18-11-10(13(21)19-15)9(12(20)17-11)6-3-7-1-4-8(5-2-7)14(22)23/h1-2,4-5,9H,3,6H2,(H,22,23)(H4,16,17,18,19,20,21)
LEDER C1=C(C=CC(=C1)C(=O)O)CCC2C3=C(N=C2O)NC(=N)N=C3O
IUPAC-namn 4-[2-(2-amino-4,6-dioxo-5,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid
Molekylvikt (g/mol) 314.3
Formel vikt 314.1015