Läs mer
Oxo Thiamine, TRC
Produktkod. 30425954
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30425954 250 mg
1 options
Produktkod. 30425954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO864200250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-2(3H)-one
CAS 490-82-4
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H16 N4 O2 S
Kvantitet 250 mg
Synonym Thiamine Hydrochloride Imp. D (EP), Thiamine Imp. D (EP), 4-Thiazolin-2-one','3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl- (6CI,8CI), 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-2(3H)-thiazolone, Thiamin thiazolone, Thiamine Hydrochloride Impurity D, Thiamine Impurity D
InChI formel InChI=1S/C12H16N4O2S/c1-7-10(3-4-17)19-12(18)16(7)6-9-5-14-8(2)15-11(9)13/h5,17H,3-4,6H2,1-2H3,(H2,13,14,15)
LEDER CC1=C(CCO)SC(=O)N1Cc2cnc(C)nc2N
IUPAC-namn 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-2-one
Molekylvikt (g/mol) 280.35
Formel vikt 280.346