Läs mer
Oxohongdenafil, TRC
Produktkod. 30417609
Change view
Kvantitet:
5 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417609 50 mg
30409238 5 mg
30409833 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417609 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84714050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1446144-70-2
Molekylformel C25 H32 N6 O4
Molekylvikt (g/mol) 480.5594
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[2-(4-ethyl-3-oxo-1-piperazinyl)acetyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Oxohongdenafil, Oxoacetildenafil
IUPAC-namn 5-[2-ethoxy-5-[2-(4-ethyl-3-oxopiperazin-1-yl)acetyl]phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)CN4CCN(CC)C(=O)C4
Specifikationer
Kemiskt namn eller material Oxohongdenafil
Kvantitet 50 mg
InChI formel InChI=1S/C25H32N6O4/c1-5-8-18-22-23(29(4)28-18)25(34)27-24(26-22)17-13-16(9-10-20(17)35-7-3)19(32)14-30-11-12-31(6-2)21(33)15-30/h9-10,13H,5-8,11-12,14-15H2,1-4H3,(H,26,27,34)
Formel vikt 480.2485