Learn More
Thermo Scientific™ Oxoid™ Anaerobic System Schrader Valve Core
Oxoid Anaerobic System Schrader Valve Core is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Brand: Thermo Scientific™ HP0022A
362.90 SEK valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Oxoid Anaerobic System Schrader Valve Core is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Specifications
Anaerobic System Schrader Valve Core | |
Thermo Scientific Oxoid | |
10/Pk. |