Learn More
Thermo Scientific™ Brucella Medium Base (Dehydrated)
Isolate and cultivate Brucella with this medium when used with Brucella Selective Supplement (SR0083 or SR0209).
Brand: Thermo Scientific™ CM0169B
Additional Details : Weight : 0.50000kg
Description
Isolate and cultivate Brucella using Thermo Scientific™ Oxoid™ Brucella Medium Base (Dehydrated) when used with Brucella Selective Supplement (SR0083 or SR0209). Brucella medium Base may be used to prepare the serum-dextrose-antibiotic medium as described by Jones and Brinley Morgan1 for the cultivation and isolation of Brucella, including fastidious types.
Isolate and cultivate Brucella using Brucella Medium Base when used with Brucella Selective Supplement (SR0083 or SR0209).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Brucella Medium Base | |
Agar | |
For 11.1L medium |
Powder | |
500 g |
For Laboratory Use Only