Läs mer
Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement, for 100mL medium
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Brand: Thermo Scientific™ SR0118E
Beskrivning
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Add to Legionella CYE Agar Base, Part No. CM0655B