Learn More
Thermo Scientific™ Oxoid™ Modified Listeria Selective Enrichment Supplement
The Oxoid Listeria Selective Supplement, Modified (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Brand: Thermo Scientific™ SR0213E
Additional Details : Weight : 0.17500kg
Description
Oxoid Listeria Selective Supplement, (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Add to to Buffered Listeria Enrichment Broth, Part No. CM0897R or Listeria Enrichment Broth, Part No. CM0862B. For the enrichment of Listeria monocytogenes. Each vial supplements 500mL of medium.Specifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Selective Supplement, Modified | |
Selective Supplement | |
Each vial for 500mL medium |