Learn More
Thermo Scientific™ Oxoid™ Modified Listeria Selective Enrichment Supplement
The Oxoid Listeria Selective Supplement, Modified (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Brand:  Thermo Scientific™ SR0213E
Additional Details : Weight : 0.17500kg
Product Code. 13255419
  • 940.00 SEK / Pack of 10
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
Description
Description
Oxoid Listeria Selective Supplement, (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Add to to Buffered Listeria Enrichment Broth, Part No. CM0897R or Listeria Enrichment Broth, Part No. CM0862B. For the enrichment of Listeria monocytogenes. Each vial supplements 500mL of medium.
Specifications
Specifications
Listeria
Freeze-dried
10/Pk.
Listeria Selective Supplement, Modified
Selective Supplement
Each vial for 500mL medium