Läs mer
Oxprenolol Hydrochloride, LoGiCal
Produktkod. 30459916
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30459916 10 mg
1 options
Produktkod. 30459916 Leverantör LoGiCal Leverantörsnummer LGCFOR0129.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxprenolol hydrochloride
CAS 6452-73-9
Analyt- eller komponentnamn Oxprenolol hydrochloride
Frakt skick Room Temperature
Molekylformel C15 H23 N O3 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yloxy)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyloxy)phenoxy]-, hydrochloride (9CI), 2-Propanol, 1-[o-(allyloxy)phenoxy]-3-(isopropylamino)-, hydrochloride (7CI,8CI), 1-(2-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(2'-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propanol hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propranol-hydrochloride, 1-Isopropyl-amino-2-hydroxy-3-(O-allyloxyphenoxy)propane hydrochloride, 1-Isopropylamino-3-(o-allyloxyphenoxy)-2-propanol hydrochloride, Ba 39089, CIBA 39089Ba, Laracor, Oxiprenolol hydrochloride, Oxprenolol hydrochloride, Paritane, Slow-Pren, Slow-Trasicor, Trasacor, Trasicor, Trasicor 80, dl-Alprenolol hydrochloride, dl-Oxprenolol hydrochloride, (+/-)-1-[o-(Allyloxy)phenoxy]-3-isopropylamino-2-propanol hydrochloride, (+/-)-Ba 39089, (+/-)-Oxprenolol hydrochloride
InChI formel InChI=1S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17H,1,9-11H2,2-3H3;1H
Lösningstyp Neat
LEDER Cl.CC(C)NCC(O)COc1ccccc1OCC=C
IUPAC-namn 1-(propan-2-ylamino)-3-(2-prop-2-enoxyphenoxy)propan-2-ol;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 301.81
Kvantitet 10 mg
Formel vikt 301.1445 g/mol