Läs mer
Oxprenolol Hydrochloride, TRC
Produktkod. 18042400
Change view
Kvantitet:
100 mg
250 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18042400 2.5 g 2.5g
18062900 100 mg -
18091210 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18042400 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8705002.5G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6452-73-9
Molekylformel C15 H23 N O3 . Cl H
Molekylvikt (g/mol) 301.81
Synonym 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yloxy)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyloxy)phenoxy]-, hydrochloride (9CI), 2-Propanol, 1-[o-(allyloxy)phenoxy]-3-(isopropylamino)-, hydrochloride (7CI,8CI), (±)-1-[o-(Allyloxy)phenoxy]-3-isopropylamino-2-propanol hydrochloride, (±)-Ba 39089, (±)-Oxprenolol hydrochloride, 1-(2-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(2'-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propanol hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propranol-hydrochloride, 1-Isopropyl-amino-2-hydroxy-3-(O-allyloxyphenoxy)propane hydrochloride, 1-Isopropylamino-3-(o-allyloxyphenoxy)-2-propanol hydrochloride, Ba 39089, CIBA 39089Ba, Laracor, Oxiprenolol hydrochloride, Oxprenolol hydrochloride, Paritane, Slow-Pren, Slow-Trasicor, Trasacor, Trasicor, Trasicor 80, dl-Alprenolol hydrochloride, dl-Oxprenolol hydrochloride
IUPAC-namn 1-(propan-2-ylamino)-3-(2-prop-2-enoxyphenoxy)propan-2-ol;hydrochloride
LEDER Cl.CC(C)NCC(O)COc1ccccc1OCC=C
Specifikationer
Kemiskt namn eller material Oxprenolol hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17H,1,9-11H2,2-3H3;1H
Formel vikt 301.1445
Procent renhet >95%