Läs mer
Oxybuprocaine Hydrochloride, LoGiCal
Produktkod. 30459844
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30459844 10 mg
1 options
Produktkod. 30459844 Leverantör LoGiCal Leverantörsnummer LGCFOR0543.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxybuprocaine Hydrochloride
CAS 5987-82-6
Analyt- eller komponentnamn Oxybuprocaine Hydrochloride
Frakt skick Room Temperature
Molekylformel C17 H28 N2 O3 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI,9CI), 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, Benoxil, Benoxinate hydrochloride, Boxinate, Cebesine, Conjucaine, Conjuncain, Dorsacaine, Dorsacaine hydrochloride, Lacrimin, Novesin, Novesine, Oxybuprocaine hydrochloride
InChI formel InChI=1S/C17H28N2O3.ClH/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3;/h8-9,13H,4-7,10-12,18H2,1-3H3;1H
Lösningstyp Neat
LEDER Cl.CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC
IUPAC-namn 2-(diethylamino)ethyl 4-amino-3-butoxybenzoate;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 344.88
Kvantitet 10 mg
Formel vikt 344.1867 g/mol