Learn More
Oxybutynin chloride, MedChemExpress
Product Code. 16408206
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16408206 100 mg
16482076 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16408206 Supplier Medchem Express Supplier No. HYB0267A
MedChemExpress Oxybutynin chloride is an anticholinergic agent, which inhibits vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM.
Oxybutynin chloride is an anticholinergic agent, which inhibits vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM.
Specifications
Kemiskt namn eller material Oxybutynin chloride
Fysisk form Solid
CAS 1508-65-2
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H32ClNO3
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : ≥ 100 mg/mL (253.84 mM) ∣H2O : 33.33 mg/mL (84.60 mM; Need ultrasonic)
LEDER O=C(C(O)(C1=CC=CC=C1)C2CCCCC2)OCC#CCN(CC)CC.[H]Cl
Molekylvikt (g/mol) 393.95
Formel vikt 393.95
Procent renhet 98.31%