Läs mer
Oxybutynin Chloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Oxybutynin Hydrochloride |
| CAS | 1508-65-2 |
| Molekylformel | C22 H31 N O3 . Cl H |
| Kvantitet | 25 g |
| Synonym | Oxybutynin Hydrochloride, Benzeneacetic acid','α-cyclohexyl-α-hydroxy-','4-(diethylamino)-2-butynyl ester','hydrochloride (9CI), Cyclohexaneglycolic acid','α-phenyl-','4-(diethylamino)-2-butynyl ester','hydrochloride (7CI,8CI), 2-Butyn-1-ol','4-(diethylamino)-','α-phenylcyclohexaneglycolate (ester)','hydrochloride (8CI), 4-(Diethylamino)-2-butynyl α-phenylcyclohexaneglycolate hydrochloride, Cystrin, Dridase, MJ 4309-1, MJ 50-58, Oxybutynin chloride, Oxybutynin hydrochloride, Pollakisu, Tropax |
| InChI formel | InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H |
| LEDER | Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC-namn | 4-(diethylamino)but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride |
| Molekylvikt (g/mol) | 393.95 |
| Formel vikt | 393.2071 |