Läs mer
Oxycodone Impurity EP C (as Hydrochloride), Mikromol™
Beskrivning
Oxycodone Impurity EP C (as Hydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Noroxycodone HCl |
| CAS | 52446-25-0 |
| Analyt- eller komponentnamn | Noroxycodone HCl |
| Frakt skick | Room Temperature |
| Molekylformel | C17 H19 N O4 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 4,5α-Epoxy-14-hydroxy-3-methoxymorphinan-6-one Hydrochloride (Noroxycodone Hydrochloride), Oxycodone Hydrochloride Imp. C (EP), Oxycodone Imp. C (EP), 4,5alpha-Epoxy-14-hydroxy-3-methoxymorphinan-6-one Hydrochloride, Oxycodone Hydrochloride Imp. C (EP) as Hydrochloride, Noroxycodone Hydrochloride, Oxycodone Hydrochloride Impurity C as Hydrochloride, Oxycodone Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C17H19NO4.ClH/c1-21-11-3-2-9-8-12-17(20)5-4-10(19)15-16(17,6-7-18-12)13(9)14(11)22-15;/h2-3,12,15,18,20H,4-8H2,1H3;1H/t12-,15+,16+,17-;/m1./s1 |
| LEDER | Cl.COc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O |
| IUPAC-namn | (4R,4aS,7aR,12bS)-4a-hydroxy-9-methoxy-1,2,3,4,5,6,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride |
| Visa mer |