Läs mer
Oxyfedrine, TRC
Produktkod. 30410813
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410813 5 mg
30420728 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30410813 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8707005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15687-41-9
Molekylformel C19 H23 N O3
Molekylvikt (g/mol) 313.39
Synonym 3-[[(1S,2R)-2-Hydroxy-1-methyl-2-phenylethyl]amino]-1-(3-methoxyphenyl)-1-propanone
IUPAC-namn 3-[[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]amino]-1-(3-methoxyphenyl)propan-1-one
LEDER COc1cccc(c1)C(=O)CCN[C@@H](C)[C@H](O)c2ccccc2
Specifikationer
Kemiskt namn eller material Oxyfedrine
Kvantitet 5 mg
InChI formel InChI=1S/C19H23NO3/c1-14(19(22)15-7-4-3-5-8-15)20-12-11-18(21)16-9-6-10-17(13-16)23-2/h3-10,13-14,19-20,22H,11-12H2,1-2H3/t14-,19-/m0/s1
Formel vikt 313.1678