Läs mer
Oxymatrine, TRC
Produktkod. 17913445
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17913445 250 mg
17931745 100 mg
17960615 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17913445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO876300250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16837-52-8
Molekylformel C15 H24 N2 O2
Molekylvikt (g/mol) 264.36
Synonym 1H,5H,10H-Dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, dodecahydro-, 4-oxide, (4R,7aS,13aR,13bR,13cS)-, Matridin-15-one, 1-oxide, (1β)-, Matrine, 1-oxide (8CI), Matrine, N-oxide (6CI,7CI), (+)-Matrine N-oxide, (+)-Oxymatrine, 1H,5H,10H-Dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, dodecahydro-, 4-oxide, [4R-(4α,7aβ,13aα,13bβ,13cβ)]-, Ammothamnine, Matrine 1β-oxide, Matrine N1-oxide, Oxymatrine
LEDER [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23
Specifikationer
Kemiskt namn eller material Oxymatrine
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C15H24N2O2/c18-14-7-1-6-13-12-5-3-9-17(19)8-2-4-11(15(12)17)10-16(13)14/h11-13,15H,1-10H2/t11-,12+,13+,15-,17?/m0/s1
Formel vikt 264.1838
Procent renhet >95%