missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Oxymetazolinhydroklorid, Mikromol™
Description
Oxymetazolin hydroklorid
Specifications
Specifications
| Kemiskt namn eller material | Oxymetazoline Hydrochloride |
| CAS | 2-8-2315 |
| Analyt- eller komponentnamn | Oxymetazoline Hydrochloride |
| Frakt skick | Rumstemperatur |
| Molekylformel | C16 H24 N2 O . Cl H |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Oxymetazoline Hydrochloride, 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol hydrochloride (1:1), 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol monohydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol monohydrochloride, 2,6-Dimethyl-4-tertiarybutyl-3-hydroxyphenyl)methylimidazoline hydrochloride, 2-(4-tert-Butyl-2,6-dimethyl-3-hydroxybenzyl)-2-imidazolinium chloride, 4-Way Nasal 12 Hour Spray, 4-Way Nasal Spray, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrazine, Afrin, Afrin hydrochloride, Allerest 12 Hour Nasal Spray, Dristan Long-Lasting Nasal Mist, Duration, Duration 12 Hour Nasal Spray, H 990, Iliadin, Nafrine, Nasivin, Neo-Synephrine 12 Hour, Neo-Synephrine 12 Hour NTZ, Nostrilla, Ocuclear, Oxilin, Oxymetazoline chloride, Oxymetazoline hydrochloride, Respir, Sch 9384 |
| InChI formel | InChI=1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H |
| LEDER | Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C |
| IUPAC-namn | 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmetyl)-2,4-dimetylfenol; hydroklorid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?