Läs mer
Oxymetholone, Lipomed
Produktkod. 30463468
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30463468 50 mg
1 options
Produktkod. 30463468 Leverantör Lipomed Leverantörsnummer LPMOXY167850
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Oxymetholone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxymetholone
CAS 434-07-1
Analyt- eller komponentnamn Oxymetholone
Frakt skick Room Temperature
Molekylformel C21 H32 O3
Synonym 17-Beta-Hydroxy-2-hydroxymethylene-17-alpha-methyl-3-androstanone, 2-(Hydroxymethylene)-17-methyldihydrotestosterone, Adroyd, Anadrol, Anapolan 50, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER C[C@]1(O)CC[C@H]2 [C@@H]3 CC[C@H]4 CC(=O)\C(=C/O)\C[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC-namn (2 Z,5 S,8 R,9 S,10 S,13 S,14 S,17 S)-17-hydroxy-2-(hydroxymethylidene)-10,13,17-trimethyl-1,4,5,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Industry Type Pharmaceutical
Molekylvikt (g/mol) 332.48
Kvantitet 50 mg
Formel vikt 332.2351 g/mol