Läs mer
Invitrogen™ Oxytocin Competitive ELISA Kit
Beskrivning
The Oxytocin (OT) ELISA quantitates OT in serum, plasma and other biological fluids. Principle of the method The Competitive Oxytocin (OT) ELISA research use only kit is designed to quantitatively measure OT independent of species. A OT standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to OT is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of oxytocin in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Specifikationer
Specifikationer
| Analysintervall | 15.63-1000 pg/mL |
| Analyskänslighet | 9.38 pg/mL |
| Konjugera | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Produkttyp | ELISA |
| Provtyp | Cell Lysates, Plasma, Serum, Tissue Homogenates |
| Målarter | All Species |
| För användning med (utrustning) | Absorbance Microplate Reader |
| Gene Alias | neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein productneurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product; |
| Interassay CV | <10% |
| Visa mer |