missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Oxytocin Polyclonal Antibody
GREENER_CHOICE

Produktkod. 17857822
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17857822 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17857822 Leverantör Invitrogen™ Leverantörsnummer PA5142427

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.

There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Oxytocin
Användningsområden Immunohistochemistry (PFA fixed)
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gen OXT
Genaccessionsnr. P01178
Gene Alias neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product
Gensymboler OXT
Värd art Goat
Immunogen Peptide with sequence HADPACDAEATFSQR (aa 111-125).
Reningsmetod Ammonium sulfate precipitation
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 5020
Målarter Human
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.