Läs mer
P-[(1R,2R)-1,2-Dihydroxypropyl]-phosphonic Acid Ammonium Salt, TRC
Produktkod. 30135692
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135692 250 mg
30003669 500 mg
30009615 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135692 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD454560250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1160525-87-0
Molekylformel C3 H9 O5 P . 2 H3 N
Molekylvikt (g/mol) 190.14
IUPAC-namn azane;[(1R,2R)-1,2-dihydroxypropyl]phosphonic acid
LEDER N.N.C[C@@H](O)[C@H](O)P(=O)(O)O
Specifikationer
Kemiskt namn eller material P-[(1R,2R)-1,2-Dihydroxypropyl]-phosphonic Acid Ammonium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C3H9O5P.2H3N/c1-2(4)3(5)9(6,7)8;;/h2-5H,1H3,(H2,6,7,8);2*1H3/t2-,3-;;/m1../s1
Formel vikt 190.0719
Procent renhet >95%