Läs mer
P-[(3-Bromo-7-cyano-2-naphthalenyl)difluoromethyl]phosphonic Acid Diammonium Salt, TRC
Produktkod. 30339163
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30339163 1 mg
1 options
Produktkod. 30339163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6827351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material P-[(3-Bromo-7-cyano-2-naphthalenyl)difluoromethyl]phosphonic Acid Diammonium Salt
CAS 809272-64-8
Molekylformel C12H13BrF2N3O3P
Kvantitet 1 mg
Synonym [(3-Bromo-7-cyano-2-naphthalenyl)difluoromethyl]phosphonic Acid Diammonium Salt
InChI formel InChI=1S/C12H7BrF2NO3P/c13-11-5-8-2-1-7(6-16)3-9(8)4-10(11)12(14,15)20(17,18)19/h1-5H,(H2,17,18,19)
LEDER OP(=O)(O)C(F)(F)c1cc2cc(ccc2cc1Br)C#N
IUPAC-namn [(3-bromo-7-cyanonaphthalen-2-yl)-difluoromethyl]phosphonic acid
Molekylvikt (g/mol) 396.12
Formel vikt 360.9315