Learn More
Welch™ P 8 E Rotary Vacuum Pump
Brand: Welch™ 321004
Additional Details : Weight : 25.00000kg
Specifications
120/143 L/min. | |
150 mm | |
<49 dB | |
21 kg | |
215 mm | |
Rotary Vane Vacuum Pump |
100/115/230 V 50/60 Hz | |
P 8 E | |
200 w | |
50/60 Hz | |
405 mm | |
100/115/230 V |