Läs mer
p-Aminophenyl Arsenoxide, TRC
Produktkod. 30323151
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30323151 5 mg
1 options
Produktkod. 30323151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6225005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Aminophenyl Arsenoxide
CAS 1122-90-3
Molekylformel C6 H6 As N O
Kvantitet 5 mg
Synonym Benzenamine, 4-arsenoso-, Aniline, p-arsenoso- (6CI,7CI,8CI), 4-Arsenosobenzenamine, (4-Aminophenyl)arsine oxide, (p-Aminophenyl)arsenoxide, NSC 10876, p-Arsenosoaniline, p-Aminophenyl arsenoxide
InChI formel InChI=1S/C6H6AsNO/c8-6-3-1-5(7-9)2-4-6/h1-4H,8H2
LEDER Nc1ccc(cc1)[As]=O
IUPAC-namn 4-arsorosoaniline
Molekylvikt (g/mol) 183.04
Formel vikt 182.9665