Läs mer
p-Azidoacetophenone, TRC
Produktkod. 30341382
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30341382 5 g
30338674 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30341382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA8212505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Azidoacetophenone
CAS 20062-24-2
Molekylformel C8 H7 N3 O
Kvantitet 5 g
InChI formel InChI=1S/C8H7N3O/c1-6(12)7-2-4-8(5-3-7)10-11-9/h2-5H,1H3
LEDER CC(=O)c1ccc(cc1)N=[N+]=[N-]
IUPAC-namn 1-(4-azidophenyl)ethanone
Molekylvikt (g/mol) 161.16
Formel vikt 161.0589