Läs mer
p-Azidobenzoylhydrazide, TRC
Produktkod. 30415160
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415160 25 mg
30414358 50 mg
30413185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29029025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63296-32-2
Molekylformel C7H7N5O
Molekylvikt (g/mol) 177.16
IUPAC-namn 4-azidobenzohydrazide
LEDER NNC(=O)c1ccc(cc1)N=[N+]=[N-]
Specifikationer
Kemiskt namn eller material p-Azidobenzoylhydrazide
Kvantitet 25 mg
InChI formel InChI=1S/C7H7N5O/c8-10-7(13)5-1-3-6(4-2-5)11-12-9/h1-4H,8H2,(H,10,13)
Formel vikt 177.0651