Läs mer
p-Chlorobenzylbenzene, TRC
Produktkod. 30353548
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30353548 25 g
1 options
Produktkod. 30353548 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36472525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Chlorobenzylbenzene
CAS 831-81-2
Molekylformel C13H11Cl
Kvantitet 25 g
Synonym 1-Chloro-4-(phenylmethyl)benzene, (p-chlorophenyl)phenylmethane, (4-Chlorophenyl)phenylmethane, (p-Chlorophenyl)phenylmethane, 1-Benzyl-4-chlorobenzene, 1-Chloro-4-benzylbenzene, 4-Benzylchlorobenzene, 4-Chlorodiphenylmethane, NSC 83166, Phenyl(p-chlorophenyl)methane, p-Chlorodiphenylmethane
InChI formel InChI=1S/C13H11Cl/c14-13-8-6-12(7-9-13)10-11-4-2-1-3-5-11/h1-9H,10H2
LEDER Clc1ccc(Cc2ccccc2)cc1
IUPAC-namn 1-benzyl-4-chlorobenzene
Molekylvikt (g/mol) 202.68
Formel vikt 202.0549