Läs mer
p-Chlorophenylguanidine Hydrochloride, TRC
Produktkod. 30364858
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30364858 250 mg
1 options
Produktkod. 30364858 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC377740250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Chlorophenyl)guanidine Hydrochloride
CAS 14279-91-5
Molekylformel C7 H8 Cl N3 . Cl H
Kvantitet 250 mg
Synonym (4-Chlorophenyl)guanidine Hydrochloride, Guanidine, N-(4-chlorophenyl)-, hydrochloride (1:1), Guanidine, (4-chlorophenyl)-, monohydrochloride (9CI), Guanidine, (p-chlorophenyl)-, monohydrochloride (8CI), 4-Chlorophenylguanidine hydrochloride, BW 65-100, p-Chlorophenyl urea hydrochloride
InChI formel InChI=1S/C7H8ClN3.ClH/c8-5-1-3-6(4-2-5)11-7(9)10;/h1-4H,(H4,9,10,11);1H
LEDER Cl.NC(=N)Nc1ccc(Cl)cc1
IUPAC-namn 1-(4-chlorophenyl)guanidine;hydrochloride
Molekylvikt (g/mol) 206.07
Formel vikt 205.0174