Läs mer
p-Cresol Rutinoside, TRC
Produktkod. 30007899
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007899 1 mg
30005164 5 mg
30003241 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007899 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7819801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1715947-02-6
Molekylformel C19 H28 O10
Molekylvikt (g/mol) 416.4196
IUPAC-namn (2S,3R,4R,5R,6R)-2-methyl-6-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methylphenoxy)oxan-2-yl]methoxy]oxane-3,4,5-triol
LEDER C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3ccc(C)cc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material p-Cresol Rutinoside
Kvantitet 1 mg
InChI formel InChI=1S/C19H28O10/c1-8-3-5-10(6-4-8)28-19-17(25)15(23)13(21)11(29-19)7-26-18-16(24)14(22)12(20)9(2)27-18/h3-6,9,11-25H,7H2,1-2H3/t9-,11+,12-,13+,14+,15-,16+,17+,18+,19+/m0/s1
Formel vikt 416.1682